Search
Search

SEL1L| Gene Symbol: SEL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105925.4 | XP_001105925.2 | 633 | 80 | ||
| GenBank mRNA | JU321757.1 | - | 612 | 80 | ||
| JU471592.1 | - | 613 | 80 | |||
| JV703743.1 | - | 607 | 80 | |||
Set Membership: |
|