Search
Search

SCN9A| Gene Symbol: SCN9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110280.2 | XP_014965766.2 | 2441 | 94 | ||
| XM_028830805.1 | XP_028686638.1 | 2261 | 94 | |||
| XM_028830806.1 | XP_028686639.1 | 2408 | 94 | |||
| XM_028830807.1 | XP_028686640.1 | 2441 | 94 | |||
Set Membership: |
|