Search
Search

MTCH1| Gene Symbol: MTCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116955.4 | XP_001116955.1 | 692 | 84 | ||
| XM_015136258.2 | XP_014991744.1 | 692 | 84 | |||
| XM_028847616.1 | XP_028703449.1 | 130 | ||||
| GenBank mRNA | JU327066.1 | - | 313 | 84 | ||
| JV678897.1 | - | 314 | 84 | |||
| JV711093.1 | - | 315 | 84 | |||
Set Membership: |
|