Search
Search

VPS4B| Gene Symbol: VPS4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257678.1 | NP_001244607.1 | 391 | 99 | ||
| GenBank mRNA | JU320343.1 | - | 391 | 99 | ||
| JU320344.1 | - | 391 | 99 | |||
| JU320345.1 | - | 391 | 99 | |||
| JU320346.1 | - | 391 | 99 | |||
| JU320347.1 | - | 391 | 99 | |||
| JU320348.1 | - | 391 | 99 | |||
| JU471316.1 | - | 482 | 99 | |||
| JV043341.1 | - | 447 | 99 | |||
Set Membership: |
|