Search
Search

KIF15| Gene Symbol: KIF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114881.4 | XP_001114881.2 | 3091 | 69 | ||
| XM_028844027.1 | XP_028699860.1 | 3091 | 69 | |||
| GenBank mRNA | JU472702.1 | - | 3022 | 69 | ||
| JU476477.1 | - | 3022 | 69 | |||
Set Membership: |
|