Search
Search

TAB3| Gene Symbol: TAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127137.2 | XP_014982623.1 | 797 | 60 | ||
| XM_015127139.2 | XP_014982625.1 | 541 | 60 | |||
| XM_015127141.2 | XP_014982627.1 | 799 | 60 | |||
| XM_028842401.1 | XP_028698234.1 | 497 | 60 | |||
| XM_028842402.1 | XP_028698235.1 | 916 | 60 | |||
| XM_028842403.1 | XP_028698236.1 | 800 | 60 | |||
| GenBank mRNA | JU333833.1 | - | 505 | 60 | ||
| JU475042.1 | - | 458 | 60 | |||
| JV044785.1 | - | 570 | 60 | |||