Search
Search

PLD2| Gene Symbol: PLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118334.2 | XP_014973820.1 | 823 | 76 | ||
| XM_028835923.1 | XP_028691756.1 | 901 | 76 | |||
| GenBank mRNA | JV636220.1 | - | 756 | 76 | ||
Set Membership: |
|