Search
Search

HELZ2| Gene Symbol: HELZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148601.2 | XP_015004087.2 | 8403 | 64 | ||
| XM_015148602.2 | XP_015004088.2 | 8455 | 64 | |||
| GenBank mRNA | JU477585.1 | - | 8379 | 64 | ||
Set Membership: |
|