Search
Search

TAGAP| Gene Symbol: TAGAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137624.2 | XP_014993110.2 | 474 | 69 | ||
| XM_028847011.1 | XP_028702844.1 | 484 | 69 | |||
| GenBank mRNA | JU474248.1 | - | 315 | 69 | ||
Set Membership: |
|