Search
Search

DGKE| Gene Symbol: DGKE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119182.2 | XP_014974668.1 | 923 | 88 | ||
| XM_028836313.1 | XP_028692146.1 | 1149 | 88 | |||
| XM_028836314.1 | XP_028692147.1 | 1151 | 88 | |||
| GenBank mRNA | JU321959.1 | - | 836 | 88 | ||
| JU473567.1 | - | 912 | 88 | |||
Set Membership: |
|