Search
Search

CNOT9| Gene Symbol: CNOT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111059.2 | XP_014966545.1 | 699 | 93 | ||
| XM_015111060.2 | XP_014966546.1 | 699 | 93 | |||
| XM_015111061.2 | XP_014966547.1 | 431 | 93 | |||
| GenBank mRNA | JU330992.1 | - | 319 | 93 | ||
| JU477771.1 | - | 329 | 93 | |||
| JV665243.1 | - | 330 | 93 | |||