Search
Search

MYOM3| Gene Symbol: MYOM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028838163.1 | XP_028693996.1 | 1620 | 93 | ||
| XM_028838166.1 | XP_028693999.1 | 1604 | 93 | |||
| GenBank mRNA | JV635584.1 | - | 1589 | 93 | ||
Set Membership: |
|