Search
Search

TRIM33| Gene Symbol: TRIM33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099267.4 | XP_001099267.4 | 652 | 60 | ||
| XM_002801713.3 | XP_002801759.3 | 652 | 60 | |||
| XM_015148754.2 | XP_015004240.2 | 652 | 60 | |||
| XM_015148762.2 | XP_015004248.2 | 652 | 60 | |||
| GenBank mRNA | JU327018.1 | - | 286 | 60 | ||
| JU327034.1 | - | 286 | 60 | |||
| JU475937.1 | - | 582 | 60 | |||
| JU475938.1 | - | 582 | 60 | |||
| JV653465.1 | - | 588 | 60 | |||
| JV716679.1 | - | 409 | 60 | |||
Set Membership: |
|