Search
Search

NFATC4| Gene Symbol: NFATC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143461.2 | XP_014998947.1 | 1439 | 100 | ||
| XM_015143462.2 | XP_014998948.1 | 1473 | 100 | |||
| XM_028851459.1 | XP_028707292.1 | 1517 | 100 | |||
| XM_028851460.1 | XP_028707293.1 | 1227 | 100 | |||
| GenBank mRNA | JV652403.1 | - | 1380 | 100 | ||
Set Membership: |
|