Search
Search

LAS1L| Gene Symbol: LAS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127590.2 | XP_014983076.1 | 322 | 59 | ||
| XM_015127591.2 | XP_014983077.1 | 322 | 59 | |||
| XM_015127592.1 | XP_014983078.1 | 327 | 59 | |||
| GenBank mRNA | JU336571.1 | - | 305 | 59 | ||
| JU336572.1 | - | 305 | 59 | |||
| JU477380.1 | - | 305 | 59 | |||
| JU477381.1 | - | 305 | 59 | |||
| JV048361.1 | - | 324 | 59 | |||
| JV048362.1 | - | 324 | 59 | |||
Set Membership: |
|