Search
Search

BACE1| Gene Symbol: BACE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260928.1 | NP_001247857.1 | 1394 | 61 | ||
| XM_015115706.2 | XP_014971192.1 | 1480 | 61 | |||
| XM_015115707.2 | XP_014971193.1 | 1423 | 61 | |||
| GenBank mRNA | JU319947.1 | - | 1262 | 61 | ||
| JU319948.1 | - | 1319 | 61 | |||
| JU319949.1 | - | 1394 | 61 | |||
| JV703565.1 | - | 1377 | 61 | |||
Set Membership: |
|