Search
Search

PPP1R21| Gene Symbol: PPP1R21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112318.2 | XP_014967804.1 | 1266 | 151 | ||
| XM_015112321.2 | XP_014967807.1 | 1302 | 151 | |||
| XM_028831687.1 | XP_028687520.1 | 1115 | 151 | |||
| XM_028831688.1 | XP_028687521.1 | 1249 | 151 | |||
| XM_028831689.1 | XP_028687522.1 | 1111 | 151 | |||
| XM_028831690.1 | XP_028687523.1 | 1288 | 151 | |||
| XM_028831691.1 | XP_028687524.1 | 1055 | 151 | |||
| GenBank mRNA | JU474323.1 | - | 1156 | 151 | ||
| JV706950.1 | - | 1220 | 151 | |||
Set Membership: |
|