Search
Search

TECRL| Gene Symbol: TECRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001110088.4 | XP_001110088.1 | 919 | 102 | ||
| XM_015138621.2 | XP_014994107.1 | 919 | 102 | |||
| XM_028848555.1 | XP_028704388.1 | 919 | 102 | |||
Set Membership: |
|