Search
Search

CCNG2| Gene Symbol: CCNG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002804071.3 | XP_002804117.3 | 1491 | 91 | ||
| XM_015138466.2 | XP_014993952.2 | 1343 | 91 | |||
| XM_015138467.2 | XP_014993953.2 | 1352 | 91 | |||
| GenBank mRNA | JU321432.1 | - | 1075 | 91 | ||
| JU472227.1 | - | 1152 | 91 | |||
| JV048621.1 | - | 1171 | 91 | |||
| JV667271.1 | - | 1164 | 91 | |||
Set Membership: |
|