Search
Search

ZDHHC17| Gene Symbol: ZDHHC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260892.1 | NP_001247821.1 | 940 | 114 | ||
| XM_015152293.2 | XP_015007779.1 | 1065 | 114 | |||
| XM_015152294.2 | XP_015007780.1 | 914 | 114 | |||
| GenBank mRNA | JU320678.1 | - | 965 | 114 | ||
| JU320679.1 | - | 965 | 114 | |||
| JU320680.1 | - | 965 | 114 | |||
| JU474925.1 | - | 940 | 114 | |||
| JV601213.1 | - | 993 | 114 | |||
Set Membership: |
|