Search
Search

CANX| Gene Symbol: CANX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101337.4 | XP_001101337.2 | 859 | 133 | ||
| XM_001101429.4 | XP_001101429.2 | 918 | 133 | |||
| XM_002804666.3 | XP_002804712.2 | 1362 | 133 | |||
| XM_015141631.2 | XP_014997117.1 | 982 | 133 | |||
| XM_015141632.2 | XP_014997118.1 | 1253 | 133 | |||
| XM_015141633.2 | XP_014997119.1 | 1144 | 133 | |||
| GenBank mRNA | JU321416.1 | - | 895 | 133 | ||
| JV048365.1 | - | 831 | 133 | |||
| JV595107.1 | - | 892 | 133 | |||
Set Membership: |
|