Search
Search

CCDC57| Gene Symbol: CCDC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120636.2 | XP_014976122.2 | 1446 | 67 | ||
| XM_028837027.1 | XP_028692860.1 | 1446 | 67 | |||
| XM_028837028.1 | XP_028692861.1 | 1439 | 67 | |||
| XM_028837029.1 | XP_028692862.1 | 1446 | 67 | |||
Set Membership: |
|