Search
Search

GNPTAB| Gene Symbol: GNPTAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028829810.1 | XP_028685643.1 | 1369 | 73 | ||
| GenBank mRNA | JU322559.1 | - | 1137 | 73 | ||
| JU472571.1 | - | 1158 | 73 | |||
Set Membership: |
|