Search
Search

NEK1| Gene Symbol: NEK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083043.4 | XP_001083043.3 | 693 | 99 | ||
| XM_001083275.4 | XP_001083275.3 | 693 | 99 | |||
| XM_015139492.2 | XP_014994978.2 | 768 | 99 | |||
| XM_015139494.2 | XP_014994980.2 | 768 | 99 | |||
| XM_028848879.1 | XP_028704712.1 | 662 | 99 | |||
| XM_028848880.1 | XP_028704713.1 | 693 | 99 | |||
| XM_028848881.1 | XP_028704714.1 | 438 | 99 | |||
| XM_028848882.1 | XP_028704715.1 | 750 | 99 | |||
| XM_028848883.1 | XP_028704716.1 | 751 | 99 | |||
| XM_028848884.1 | XP_028704717.1 | 750 | 99 | |||
Set Membership: |
|