Search
Search

RBKS| Gene Symbol: RBKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100371.4 | XP_001100371.1 | 700 | 106 | ||
| XM_001100564.3 | XP_001100564.1 | 700 | 106 | |||
| XM_015111995.2 | XP_014967481.1 | 572 | 106 | |||
| XM_015111996.2 | XP_014967482.1 | 952 | 106 | |||
| XM_015111997.2 | XP_014967483.1 | 572 | 106 | |||
| XM_015111998.2 | XP_014967484.1 | 598 | 106 | |||
| XM_015111999.2 | XP_014967485.1 | 778 | 106 | |||
| XM_015112000.2 | XP_014967486.1 | 441 | 106 | |||
| XM_028831827.1 | XP_028687660.1 | 572 | 106 | |||
| GenBank mRNA | JU325961.1 | - | 436 | 106 | ||
| JV658553.1 | - | 482 | 106 | |||
Set Membership: |
|