Search
Search

SHMT2| Gene Symbol: SHMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152100.2 | XP_015007586.1 | 687 | 110 | ||
| XM_015152101.2 | XP_015007587.1 | 393 | 110 | |||
| XM_015152102.2 | XP_015007588.1 | 349 | 110 | |||
| XM_015152104.2 | XP_015007590.1 | 393 | 110 | |||
| GenBank mRNA | JU321404.1 | - | 308 | 110 | ||
| JU476819.1 | - | 292 | 110 | |||
| JV047946.1 | - | 255 | 110 | |||
| JV590905.1 | - | 333 | 110 | |||
Set Membership: |
|