Search
Search

TACR1| Gene Symbol: TACR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037861.1 | NP_001032950.1 | 877 | 62 | ||
| GenBank mRNA | AB180074.1 | - | 877 | 62 | ||
| JV703999.1 | - | 1314 | 62 | |||
Set Membership: |
|