Search
Search

CCR6| Gene Symbol: CCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032935.1 | NP_001028107.1 | 1073 | 86 | ||
| XM_015137658.2 | XP_014993144.1 | 2991 | 86 | |||
| XM_015137659.2 | XP_014993145.1 | 1435 | 86 | |||
| XM_028846613.1 | XP_028702446.1 | 1540 | 86 | |||
| XM_028846614.1 | XP_028702447.1 | 1520 | 86 | |||
| GenBank mRNA | AF508730.1 | - | 1073 | 86 | ||
| JU471420.1 | - | 1168 | 86 | |||
Set Membership: |
|