Search
Search

UTS2| Gene Symbol: UTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032895.1 | NP_001028067.1 | 257 | 109 | ||
| XM_028850853.1 | XP_028706686.1 | 306 | 109 | |||
| GenBank mRNA | AY065983.1 | - | 257 | 109 | ||
Set Membership: |
|