Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: B2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047137.1 | NP_001040602.1 | 383 | 68 | ||
GenBank mRNA | AY349163.1 | - | 383 | 68 | ||
AY445834.1 | - | 383 | 68 | |||
JV045380.1 | - | 381 | 68 | |||
JV669635.1 | - | 436 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |