Search
Search

TOP2B| Gene Symbol: TOP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265701.1 | NP_001252630.1 | 1260 | 96 | ||
| XM_015132251.2 | XP_014987737.2 | 1913 | 96 | |||
| GenBank mRNA | AY680449.1 | - | 402 | 96 | ||
| JU327214.1 | - | 1177 | 96 | |||
| JV489123.1 | - | 1260 | 96 | |||
| JV627420.1 | - | 1189 | 96 | |||
| JV694960.1 | - | 1261 | 96 | |||
Set Membership: |
|