Search
Search

ATP2B4| Gene Symbol: ATP2B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032926.1 | NP_001028098.1 | 2840 | 80 | ||
| XM_015118176.2 | XP_014973662.1 | 3855 | 80 | |||
| XM_015118192.2 | XP_014973678.1 | 3980 | 80 | |||
| XM_015118208.2 | XP_014973694.1 | 3601 | 80 | |||
| XM_015118217.2 | XP_014973703.1 | 3822 | 80 | |||
| XM_015118221.2 | XP_014973707.1 | 3857 | 80 | |||
| XM_015118226.2 | XP_014973712.1 | 3860 | 80 | |||
| XM_028851247.1 | XP_028707080.1 | 3601 | 80 | |||
| XM_028851292.1 | XP_028707125.1 | 3824 | 80 | |||
| GenBank mRNA | AY928176.1 | - | 2840 | 80 | ||
| JU321001.1 | - | 3697 | 80 | |||
| JU321486.1 | - | 3697 | 80 | |||
| JU471377.1 | - | 3702 | 80 | |||
| JV043370.1 | - | 3700 | 80 | |||
Set Membership: |
|