Search
Search

PSMA5| Gene Symbol: PSMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260609.2 | NP_001247538.1 | 111 | 98 | ||
| GenBank mRNA | JU335513.1 | - | 106 | 98 | ||
| JU476453.1 | - | 112 | 98 | |||
| JV047692.1 | - | 111 | 98 | |||
Set Membership: |
|