Search
Search

SNX3| Gene Symbol: SNX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257953.1 | NP_001244882.1 | 691 | 152 | ||
| GenBank mRNA | JU320077.1 | - | 691 | 152 | ||
| JU328562.1 | - | 157 | 152 | |||
| JU471826.1 | - | 696 | 152 | |||
| JV048574.1 | - | 723 | 152 | |||
| JV489150.1 | - | 304 | 152 | |||
| JV597111.1 | - | 711 | 152 | |||
| JV632129.1 | - | 704 | 152 | |||
Set Membership: |
|