Search
Search

BHLHE40| Gene Symbol: BHLHE40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257518.1 | NP_001244447.1 | 409 | 121 | ||
| GenBank mRNA | JU336853.1 | - | 409 | 121 | ||
| JU477597.1 | - | 408 | 121 | |||
| JV561297.1 | - | 459 | 121 | |||
Set Membership: |
|