Search
Search

ABCA8| Gene Symbol: ABCA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120199.2 | XP_014975685.2 | 4007 | 71 | ||
| GenBank mRNA | JU332481.1 | - | 3999 | 71 | ||
| JV045893.1 | - | 4001 | 71 | |||
| JV045894.1 | - | 4001 | 71 | |||
| JV045895.1 | - | 4001 | 71 | |||
Set Membership: |
|