Search
Search

MRPL38| Gene Symbol: MRPL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101855.4 | XP_001101855.2 | 1220 | 73 | ||
| GenBank mRNA | JU335974.1 | - | 773 | 73 | ||
| JU470569.1 | - | 695 | 73 | |||
| JV047968.1 | - | 712 | 73 | |||
| JV693738.1 | - | 771 | 73 | |||
Set Membership: |
|