Search
Search

SNTA1| Gene Symbol: SNTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266239.1 | NP_001253168.1 | 485 | 139 | ||
| GenBank mRNA | JU336049.1 | - | 485 | 139 | ||
| JV626662.1 | - | 551 | 139 | |||
| JV636332.1 | - | 319 | 139 | |||
| JV668065.1 | - | 505 | 139 | |||
Set Membership: |
|