Search
Search

WAC| Gene Symbol: WAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106227.4 | XP_001106227.2 | 2168 | 70 | ||
| XM_015146685.2 | XP_015002171.1 | 2172 | 70 | |||
| XM_015146686.2 | XP_015002172.1 | 2169 | 70 | |||
| XM_015146687.2 | XP_015002173.1 | 2154 | 70 | |||
| XM_015146689.2 | XP_015002175.1 | 2134 | 70 | |||
| XM_015146690.2 | XP_015002176.1 | 1863 | 70 | |||
| XM_015146691.2 | XP_015002177.1 | 1860 | 70 | |||
| XM_015146692.2 | XP_015002178.1 | 1845 | 70 | |||
| XM_028826126.1 | XP_028681959.1 | 2227 | 70 | |||
| GenBank mRNA | JU334863.1 | - | 1511 | 70 | ||
| JU334864.1 | - | 1820 | 70 | |||
| JU334865.1 | - | 1820 | 70 | |||
| JU471538.1 | - | 1526 | 70 | |||
| JU471539.1 | - | 1835 | 70 | |||
| JU475884.1 | - | 1826 | 70 | |||
| JU475885.1 | - | 1853 | 70 | |||
| JV047288.1 | - | 1517 | 70 | |||
| JV047289.1 | - | 1826 | 70 | |||
Set Membership: |
|