Search
Search

MEIS3| Gene Symbol: MEIS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124350.2 | XP_014979836.1 | 1442 | 109 | ||
| XM_028838971.1 | XP_028694804.1 | 1391 | 109 | |||
| GenBank mRNA | JU335532.1 | - | 1122 | 109 | ||
| JV047700.1 | - | 1100 | 109 | |||
| JV551432.1 | - | 1127 | 109 | |||
| JV600302.1 | - | 1245 | 109 | |||
| JV622681.1 | - | 1296 | 109 | |||
Set Membership: |
|