Search
Search

IRF3| Gene Symbol: IRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135797.1 | NP_001129269.1 | 392 | 64 | ||
| GenBank mRNA | EU204919.1 | - | 392 | 64 | ||
| JV600278.1 | - | 562 | 64 | |||
| JV650561.1 | - | 599 | 64 | |||
Set Membership: |
|