Search
Search

CDC27| Gene Symbol: CDC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119357.2 | XP_014974843.1 | 1890 | 102 | ||
| XM_015119358.2 | XP_014974844.1 | 1860 | 102 | |||
| XM_015119359.2 | XP_014974845.1 | 1833 | 102 | |||
| XM_015119361.2 | XP_014974847.1 | 1874 | 102 | |||
| XM_015119362.2 | XP_014974848.1 | 1527 | 102 | |||
| XM_028836998.1 | XP_028692831.1 | 1419 | 102 | |||
| GenBank mRNA | JU320566.1 | - | 1832 | 102 | ||
| JU320567.1 | - | 1832 | 102 | |||
| JU478102.1 | - | 1783 | 102 | |||
| JU478103.1 | - | 1801 | 102 | |||
Set Membership: |
|