Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02807389_m1
See other EIF4G3 GE Assays ›
Gene Symbol:
EIF4G3
Gene Name:
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 203529648 - 203904488 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02807389_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: EIF4G3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015129921.2
XP_014985407.2
4891
78
XM_015129934.2
XP_014985420.2
4594
78
XM_015129991.2
XP_014985477.2
4777
78
XM_015130005.2
XP_014985491.2
4744
78
XM_015130013.2
XP_014985499.2
4723
78
XM_015130022.2
XP_014985508.2
4453
78
XM_015130031.2
XP_014985517.2
4749
78
XM_015130042.2
XP_014985528.2
4698
78
XM_015130051.2
XP_014985537.2
4659
78
XM_028838445.1
XP_028694278.1
4358
78
XM_028838448.1
XP_028694281.1
4340
78
XM_028838453.1
XP_028694286.1
4328
78
XM_028838455.1
XP_028694288.1
4307
78
XM_028838457.1
XP_028694290.1
4304
78
XM_028838458.1
XP_028694291.1
4509
78
XM_028838461.1
XP_028694294.1
4250
78
XM_028838463.1
XP_028694296.1
4247
78
XM_028838467.1
XP_028694300.1
4395
78
XM_028838469.1
XP_028694302.1
4588
78
XM_028838470.1
XP_028694303.1
4450
78
XM_028838477.1
XP_028694310.1
4555
78
XM_028838481.1
XP_028694314.1
4417
78
XM_028838482.1
XP_028694315.1
4426
78
XM_028838492.1
XP_028694325.1
4444
78
XM_028838496.1
XP_028694329.1
4013
78
XM_028838498.1
XP_028694331.1
4441
78
GenBank mRNA
JU322424.1
-
4340
78
JU477629.1
-
4345
78
JV048563.1
-
4303
78
JV489126.1
-
4333
78
JV725936.1
-
4337
78
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Macaca mulatta
Panther Classification:
Molecular Function -
translation factor
translation initiation factor
translational protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline