Search
Search

ACSL5| Gene Symbol: ACSL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148186.2 | XP_015003672.2 | 1688 | 63 | ||
| XM_015148189.2 | XP_015003675.2 | 1598 | 63 | |||
| XM_015148190.2 | XP_015003676.2 | 1743 | 63 | |||
| GenBank mRNA | JU478304.1 | - | 1519 | 63 | ||
Set Membership: |
|