Search
Search

RSRC2| Gene Symbol: RSRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151762.2 | XP_015007248.1 | 884 | 70 | ||
| XM_015151763.2 | XP_015007249.1 | 884 | 70 | |||
| XM_015151764.2 | XP_015007250.1 | 965 | 70 | |||
| XM_015151765.2 | XP_015007251.1 | 805 | 70 | |||
| XM_015151766.2 | XP_015007252.1 | 983 | 70 | |||
| XM_015151768.2 | XP_015007254.1 | 805 | 70 | |||
| GenBank mRNA | JU335983.1 | - | 867 | 70 | ||
| JU476864.1 | - | 836 | 70 | |||
| JU476865.1 | - | 836 | 70 | |||
| JV047973.1 | - | 877 | 70 | |||
| JV659192.1 | - | 834 | 70 | |||
Set Membership: |
|