Search
Search

ELF2| Gene Symbol: ELF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087465.4 | XP_001087465.2 | 642 | 143 | ||
| XM_001087584.4 | XP_001087584.2 | 606 | 143 | |||
| XM_015139234.2 | XP_014994720.1 | 1014 | 143 | |||
| XM_015139236.2 | XP_014994722.1 | 1039 | 143 | |||
| XM_015139237.2 | XP_014994723.1 | 803 | 143 | |||
| XM_015139243.2 | XP_014994729.1 | 564 | 143 | |||
| XM_015139244.2 | XP_014994730.1 | 620 | 143 | |||
| XM_015139245.2 | XP_014994731.1 | 528 | 143 | |||
| XM_028848775.1 | XP_028704608.1 | 878 | 143 | |||
| XM_028848777.1 | XP_028704610.1 | 851 | 143 | |||
| XM_028848778.1 | XP_028704611.1 | 748 | 143 | |||
| GenBank mRNA | JU333295.1 | - | 702 | 143 | ||
| JU333296.1 | - | 738 | 143 | |||
| JU333297.1 | - | 577 | 143 | |||
| JU471415.1 | - | 557 | 143 | |||
| JU474609.1 | - | 740 | 143 | |||
| JU474610.1 | - | 521 | 143 | |||
| JV046389.1 | - | 585 | 143 | |||
| JV046390.1 | - | 549 | 143 | |||
| JV708928.1 | - | 742 | 143 | |||
Set Membership: |
|