Search
Search

CLDN2| Gene Symbol: CLDN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127961.2 | XP_014983447.1 | 183 | 150 | ||
| XM_015127962.2 | XP_014983448.1 | 183 | 150 | |||
| XM_028842398.1 | XP_028698231.1 | 183 | 150 | |||
Set Membership: |
|