Search
Search

RTL6| Gene Symbol: RTL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194443.3 | NP_001181372.1 | 1252 | 78 | ||
| XM_015150409.2 | XP_015005895.1 | 1598 | 78 | |||
| GenBank mRNA | JU320997.1 | - | 1297 | 78 | ||
| JU470727.1 | - | 1448 | 78 | |||
| JU470728.1 | - | 1448 | 78 | |||
| JV044501.1 | - | 1447 | 78 | |||
Set Membership: |
|