Search
Search

SLC39A3| Gene Symbol: SLC39A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190948.1 | NP_001177877.1 | 397 | 89 | ||
| GenBank mRNA | DR770001.1 | - | 401 | 89 | ||
| EG704623.1 | - | 384 | 89 | |||
| JU334228.1 | - | 410 | 89 | |||
| JU475343.1 | - | 425 | 89 | |||
| JV046892.1 | - | 429 | 89 | |||
| JV682019.1 | - | 405 | 89 | |||
Set Membership: |
|