Search
Search

GLRB| Gene Symbol: GLRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266452.1 | NP_001253381.1 | 1041 | 80 | ||
| XM_028848039.1 | XP_028703872.1 | 962 | 80 | |||
| GenBank mRNA | JU334046.1 | - | 1075 | 80 | ||
| JU475215.1 | - | 1041 | 80 | |||
| JV712313.1 | - | 1088 | 80 | |||
Set Membership: |
|